Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R16A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550096
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550096 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550096 Supplier Invitrogen™ Supplier No. PA550096
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total MYPT3 protein.

Inhibits protein phosphatase 1 activity toward phosphorylase, myosin light chain and myosin substrates. Jikuya H., Submitted (JUL-2002) to the EMBL/GenBank/DDBJ databases. Kalnine N., Submitted (MAY-2003) to the EMBL/GenBank/DDBJ databases. The MGC Project Team, Genome Res. 14:2121-2127(2004).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R16A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PPP1R16A
Gene Accession No. Q96I34
Gene Alias 2900084E10Rik; mFLJ00392 protein; myosin phosphatase targeting subunit 3; myosin phosphatase, target subunit 3; myosin phosphatase-targeting subunit 3; Mypt3; Ppp1r16a; Protein phosphatase 1 regulatory subunit 16A; protein phosphatase 1, regulatory (inhibitor) subunit 16A; protein phosphatase 1, regulatory subunit 16A; R75527
Gene Symbols PPP1R16A
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human PPP1R16A (aa 362-411).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362944, 84988
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.