Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPP1R16A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101784
Description
Antibody detects endogenous levels of total PPP1R16A.
Inhibits protein phosphatase 1 activity toward phosphorylase, myosin light chain and myosin substrates. Jikuya H., Submitted (JUL-2002) to the EMBL/GenBank/DDBJ databases. Kalnine N., Submitted (MAY-2003) to the EMBL/GenBank/DDBJ databases. The MGC Project Team, Genome Res. 14:2121-2127(2004).
Specifications
PPP1R16A | |
Polyclonal | |
Unconjugated | |
PPP1R16A | |
2900084E10Rik; mFLJ00392 protein; myosin phosphatase targeting subunit 3; myosin phosphatase, target subunit 3; myosin phosphatase-targeting subunit 3; Mypt3; Ppp1r16a; Protein phosphatase 1 regulatory subunit 16A; protein phosphatase 1, regulatory (inhibitor) subunit 16A; protein phosphatase 1, regulatory subunit 16A; R75527 | |
Rabbit | |
Affinity chromatography | |
RUO | |
362944, 84988 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96I34 | |
PPP1R16A | |
A synthesized peptide derived from human PPP1R16A(Accession Q96I34), corresponding to amino acid residues Q364-R414. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction