Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R16B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140027
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140027 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140027 Supplier Invitrogen™ Supplier No. PA5140027
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: RASLSDRTNL YRKEYEGEAI LWQRSAAEDQ RTSTYNGDIR ETRTDQENKD PNPRLEKPVL LSEFPTKI Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 96%.

The protein encoded by this gene is membrane-associated and contains five ankyrin repeats, a protein phosphatase-1-interacting domain, and a carboxy-terminal CAAX box domain. Synthesis of the encoded protein is inhibited by transforming growth factor beta-1. The protein may bind to the membrane through its CAAX box domain and may act as a signaling molecule through interaction with protein phosphatase-1. Alternatively spliced transcript variants encoding different isoforms have been identified in this gene.[provided by RefSeq, Feb 2010].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R16B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R16B
Gene Accession No. Q96T49
Gene Alias ANKRD4; ankyrin repeat domain protein 4; ankyrin repeat domain-containing protein 4; C130078N17Rik; CAAX box protein TIMAP; hTIMAP; KIAA0823; PPP1R16B; Protein phosphatase 1 regulatory inhibitor subunit 16B; protein phosphatase 1 regulatory subunit 16B; protein phosphatase 1, regulatory (inhibitor) subunit 16B; protein phosphatase 1, regulatory subunit 16B; TGF-beta-inhibited membrane-associated protein; TIMAP; Wdt4; whn-dependent transcript 4
Gene Symbols PPP1R16B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP1R16B. Recombinant protein control fragment (Product #RP-109699).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26051
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.