Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R17 Monoclonal Antibody (3E6)
GREENER_CHOICE

Catalog No. PIMA521172
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521172 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521172 Supplier Invitrogen™ Supplier No. MA521172
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MMSTEQMQPL ELSEDRLDKL DPRCSHLDDL SDQFIKDCDL KKKPRKGKNV QATLNVESDQ KKPRRKDTPA LHIPPFIPGV FSEHLIKRYD VQERHPKGK.

Inhibits phosphatase activities of protein phosphatase 1 (PP1) and protein phosphatase 2A (PP2A) complexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R17
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3E6
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PPP1R17
Gene Accession No. O96001
Gene Alias C7orf16; GSBS; G-substrate; PPP1R17; Protein phosphatase 1 regulatory subunit 17; protein phosphatase 1, regulatory subunit 17
Gene Symbols PPP1R17
Host Species Mouse
Immunogen PPP1R17 (NP_006649, 1 a.a. ~ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10842
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.