Learn More
PPP1R17 Mouse anti-Human, Clone: OTI2A8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808959

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is found primarily in cerebellar Purkinje cells, where it functions as a protein phosphatase inhibitor. The encoded protein is a substrate for cGMP-dependent protein kinase. An allele of this gene was discovered that increases susceptibility to hypercholesterolemia. Two transcript variants encoding different isoforms have been found for this gene.Specifications
PPP1R17 | |
Monoclonal | |
Unconjugated | |
PPP1R17 | |
C7orf16, GSBS | |
Mouse | |
Affinity Chromatography | |
RUO | |
10842 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2A8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O96001 | |
PPP1R17 | |
Full length human recombit protein of human C7ORF16 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.