Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584916
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584916 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584916 Supplier Invitrogen™ Supplier No. PA584916
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPFIPGVFSE HLIKRYDVQE RHPKGKMIPV LHNTDLEQKK PRRKDTPALH MS Highest antigen sequence indentity to the following orthologs: Mouse - 81%, Rat - 81%.

Inhibits phosphatase activities of protein phosphatase 1 (PP1) and protein phosphatase 2A (PP2A) complexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R17
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R17
Gene Accession No. O96001
Gene Alias C7orf16; GSBS; G-substrate; PPP1R17; Protein phosphatase 1 regulatory subunit 17; protein phosphatase 1, regulatory subunit 17
Gene Symbols PPP1R17
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP1R17. Recombinant protein control fragment (Product #RP-108608).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10842
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.