Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP1R18 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297370AF350
Description
PPP1R18 Polyclonal specifically detects PPP1R18 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PPP1R18 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
170954 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
HKMT1098, KIAA1949, phostensin, PPP1R18, Protein Phosphatase 1 F-Actin Cytoskeleton Targeting Subunit, Protein Phosphatase 1 F-Actin Cytoskeleton-Targeting Subunit, Protein Phosphatase 1 Regulatory Subunit 18, Protein Phosphatase 1, Regulatory Subunit 18 | |
Produced in rabbits immunized with E. coli-derived Human PPP1R18 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction