Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560812
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560812 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560812 Supplier Invitrogen™ Supplier No. PA560812
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSPGESAYQK LGLTEAHKWR PDSRESQEQS LVQLEATEWR LRSGEERQDY SEECGRKEEW PVPGVAPKET AELSETLTRE AQGNSSAGVE Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 57%.

PHTNS interacts with regulatory subunits that target the enzyme to different cellular locations and change its activity toward specific substrates. Phostensin is a regulatory subunit that targets PP1 to F-actin cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R18
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R18
Gene Accession No. Q6NYC8
Gene Alias HKMT1098; KIAA1949; Phostensin; PPP1R18; protein phosphatase 1 F-actin cytoskeleton targeting subunit; Protein phosphatase 1 F-actin cytoskeleton-targeting subunit; Protein phosphatase 1 regulatory subunit 18; protein phosphatase 1, regulatory subunit 18
Gene Symbols PPP1R18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP1R18. Recombinant protein control fragment (Product #RP-99603).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170954
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.