Learn More
PPP1R18 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305616AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Monkey
PHTNS interacts with regulatory subunits that target the enzyme to different cellular locations and change its activity toward specific substrates. Phostensin is a regulatory subunit that targets PP1 to F-actin cytoskeleton.Specifications
PPP1R18 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q6NYC8 | |
PPP1R18 | |
Between 563 and 613 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation | |
Unconjugated | |
PPP1R18 | |
HKMT1098, KIAA1949, phostensin, protein phosphatase 1 F-actin cytoskeleton targeting subunit, protein phosphatase 1 regulatory subunit 18, PPP1R18 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
170954 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.