Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPP1R1A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA573002
Description
PPP1R1A Polyclonal Antibody for Western Blot, IHC (P)
Protein phosphatase inhibitor-1 (also designated inhibitor of protein phospha-tase 1, IPP-1 and I-1) plays a role in regulating the phosphorylation of other proteins, and is itself phosphorylated by a cyclic AMP-dependent protein kinase at Threonine 35. In addition, the proline-directed kinases Cdk1, Cdk5, and mitogen-activated protein kinase (MAPK) mediate in vitro phosphorylation of IPP-1 at the phylogenetically conserved position Serine 67. In striatal tissues, glutamate-dependent regulation of N-methyl-d-aspartic acid-type channels influences IPP-1 phosphorylation at Ser 67. The localization and expression of IPP-1 suggests that it may play discrete roles in certain regions and developing stages of the brain, independent of the regulation of protein phosphatase type 1 (PP-1). PP-1 binds to both phosphorylated and dephosphorylated IPP-1. Conversion of PP-1 to a Mn++-dependent state appears to play a role in its regulation by IPP-1. IPP-1 attenuates the activity of glycogen phosphorylase and is thought to be important in the hormonal control of glycogen metabolism.
Specifications
PPP1R1A | |
Polyclonal | |
Unconjugated | |
Ppp1r1a | |
0610038N18Rik; I1; I-1; inhibitor-1; Ipp1; IPP-1; PP1 inhibitor 1; Ppp1r1a; protein phosphatase 1 regulatory inhibitor subunit 1A; protein phosphatase 1 regulatory subunit 1A; protein phosphatase 1, regulatory (inhibitor) subunit 1A; protein phosphatase inhibitor 1; protein phosphatase inhibitor-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100064738, 100855706, 5502 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.2% gelatin and 0.05% sodium azide | |
Q13522, Q8WMS3 | |
Ppp1r1a | |
A portion of amino acids 50-100 of human IPP-1 was used as the immunogen. | |
100 μg | |
Primary | |
Human, Canine, Monkey, Horse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction