Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102875
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102875 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102875 Supplier Invitrogen™ Supplier No. PA5102875
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PPP1R1A.

Protein phosphatase inhibitor-1 (also designated inhibitor of protein phospha-tase 1, IPP-1 and I-1) plays a role in regulating the phosphorylation of other proteins, and is itself phosphorylated by a cyclic AMP-dependent protein kinase at Threonine 35. In addition, the proline-directed kinases Cdk1, Cdk5, and mitogen-activated protein kinase (MAPK) mediate in vitro phosphorylation of IPP-1 at the phylogenetically conserved position Serine 67. In striatal tissues, glutamate-dependent regulation of N-methyl-d-aspartic acid-type channels influences IPP-1 phosphorylation at Ser 67. The localization and expression of IPP-1 suggests that it may play discrete roles in certain regions and developing stages of the brain, independent of the regulation of protein phosphatase type 1 (PP-1). PP-1 binds to both phosphorylated and dephosphorylated IPP-1. Conversion of PP-1 to a Mn++-dependent state appears to play a role in its regulation by IPP-1. IPP-1 attenuates the activity of glycogen phosphorylase and is thought to be important in the hormonal control of glycogen metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R1A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ppp1r1a
Gene Accession No. Q13522
Gene Alias 0610038N18Rik; I1; I-1; inhibitor-1; Ipp1; IPP-1; PP1 inhibitor 1; Ppp1r1a; protein phosphatase 1 regulatory inhibitor subunit 1A; protein phosphatase 1 regulatory subunit 1A; protein phosphatase 1, regulatory (inhibitor) subunit 1A; protein phosphatase inhibitor 1; protein phosphatase inhibitor-1
Gene Symbols Ppp1r1a
Host Species Rabbit
Immunogen A synthesized peptide derived from human PPP1R1A(Accession Q13522), corresponding to amino acid residues G128-V171.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5502
Target Species Human, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.