Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R1A Recombinant Rabbit Monoclonal Antibody (SN0756)
GREENER_CHOICE

Catalog No. PIMA532310
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532310 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532310 Supplier Invitrogen™ Supplier No. MA532310
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Protein phosphatase inhibitor-1 (also designated inhibitor of protein phospha-tase 1, IPP-1 and I-1) plays a role in regulating the phosphorylation of other proteins, and is itself phosphorylated by a cyclic AMP-dependent protein kinase at Threonine 35. In addition, the proline-directed kinases Cdk1, Cdk5, and mitogen-activated protein kinase (MAPK) mediate in vitro phosphorylation of IPP-1 at the phylogenetically conserved position Serine 67. In striatal tissues, glutamate-dependent regulation of N-methyl-d-aspartic acid-type channels influences IPP-1 phosphorylation at Ser 67. The localization and expression of IPP-1 suggests that it may play discrete roles in certain regions and developing stages of the brain, independent of the regulation of protein phosphatase type 1 (PP-1). PP-1 binds to both phosphorylated and dephosphorylated IPP-1. Conversion of PP-1 to a Mn++-dependent state appears to play a role in its regulation by IPP-1. IPP-1 attenuates the activity of glycogen phosphorylase and is thought to be important in the hormonal control of glycogen metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R1A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone SN0756
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% Glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene Ppp1r1a
Gene Accession No. P19103, Q13522, Q9ERT9
Gene Alias 0610038N18Rik; I1; I-1; inhibitor-1; Ipp1; IPP-1; PP1 inhibitor 1; Ppp1r1a; protein phosphatase 1 regulatory inhibitor subunit 1A; protein phosphatase 1 regulatory subunit 1A; protein phosphatase 1, regulatory (inhibitor) subunit 1A; protein phosphatase inhibitor 1; protein phosphatase inhibitor-1
Gene Symbols Ppp1r1a
Host Species Rabbit
Immunogen Synthetic peptide within Human PPP1R1A aa 1-50.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5502, 58200, 58977
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.