Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104038
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104038 Supplier Invitrogen™ Supplier No. PA5104038
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PPP1R1C.

Protein phosphatase-1.[supplied by OMIM, Feb 2010].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R1C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PPP1R1C
Gene Accession No. Q8BKK4, Q8WVI7
Gene Alias 4930565M23Rik; 4930579P15Rik; inhibitor-5 of protein phosphatase 1; IPP5; PP1 inhibitor protein 5; PPP1R1C; protein phosphatase 1 regulatory inhibitor subunit 1C; protein phosphatase 1 regulatory subunit 1A; protein phosphatase 1 regulatory subunit 1C; protein phosphatase 1, regulatory (inhibitor) subunit 1C; RGD1562095
Gene Symbols PPP1R1C
Host Species Rabbit
Immunogen A synthesized peptide derived from human PPP1R1C(Accession Q8WVI7), corresponding to amino acid residues N4-G54.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151242, 499818, 75276
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.