Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R2 (phospho S121/S122) Rabbit anti-Human, Mouse, Rat, Polyclonal , Abnova™

Catalog No. 89956781 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-956-781 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-956-781 Supplier Abnova Corporation Supplier No. PAB29586
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of human PPP1R2.

Specifications

Antigen PPP1R2 (phospho S121/S122)
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of human PPP1R2.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
Gene Alias IPP2/MGC87148
Gene Symbols PPP1R2
Host Species Rabbit
Immunogen A synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S120/S121 of human PPP1R2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5504
Test Specificity PPP1R2 (phospho S120/S121) polyclonal antibody detects endogenous levels of human PPP1R2 only when phosphorylated at serine 120 and serine 121.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.