Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP1R21 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA557820
Description
Immunogen sequence: LTRKYMETIE KLQNDKAKLE VKSQTLEKEA KECRLRTEEC QLQLKTLHED LSGRLEESLS IINEKVPFND TKYSQYNALN VPLHNRRHQL KMRDIA Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 96%.
CCDC128, coiled-coil domain containing 128, coiled-coil domain-containing protein 128, KLRAQ motif containing 1, KLRAQ motif-containing protein 1, KLRAQ1, protein phosphatase 1 regulatory subunit 21
Specifications
PPP1R21 | |
Polyclonal | |
Unconjugated | |
PPP1R21 | |
CCDC128; coiled-coil domain containing 128; coiled-coil domain-containing protein 128; KLRAQ motif containing 1; KLRAQ motif-containing protein 1; KLRAQ1; PPP1R21; Protein phosphatase 1 regulatory subunit 21; protein phosphatase 1, regulatory subunit 21 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
129285 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZMI0 | |
PPP1R21 | |
Recombinant protein corresponding to Human PPP1R21 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction