Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP1R26 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306098PCP
Description
PPP1R26 Polyclonal antibody specifically detects PPP1R26 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| PPP1R26 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| DRIM/UTP20 interacting protein, KIAA0649,1A6/DRIM (down-regulated in metastasis) interacting protein | |
| Produced in rabbits immunized with E. coli-derived Human PPP1R26 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 9858 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction