Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560794
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560794 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560794 Supplier Invitrogen™ Supplier No. PA560794
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NEKRQHETQK CDGSVEKKPD TNENSAKSLL KSHQEPPTKV VHRQGLLGVQ KEFAFRKPPR LAKMNVQPRS LRSKVTTTQE NEGSTKPATP CRPSEAAQNK G Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 51%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R26
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R26
Gene Accession No. Q5T8A7
Gene Alias 1A6/DRIM (down-regulated in metastasis) interacting protein; DRIM/UTP20 interacting protein; KIAA0649; PPP1R26; Protein phosphatase 1 regulatory subunit 26; protein phosphatase 1, regulatory subunit 26
Gene Symbols PPP1R26
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP1R26. Recombinant protein control fragment (Product #RP-100466).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9858
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.