Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R27 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139798
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139798 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5139798 Supplier Invitrogen™ Supplier No. PA5139798
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: YLISLGADRD ATNDDGDLPS DLIDPDYKEL VELFKGTTMD Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 93%.

Inhibits phosphatase activity of protein phosphatase 1 (PP1) complexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R27
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R27
Gene Accession No. Q86WC6
Gene Alias dysferlin interacting protein 1; Dysferlin-interacting protein 1; dysferlin-interacting protein 1 (toonin); DYSFIP1; PPP1R27; Protein phosphatase 1 regulatory subunit 27; protein phosphatase 1, regulatory subunit 27; toonin
Gene Symbols PPP1R27
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP1R27. Recombinant protein control fragment (Product #RP-98932).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116729
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.