Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R2P3 Monoclonal Antibody (1H2)
GREENER_CHOICE

Catalog No. PIMA523117
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523117 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523117 Supplier Invitrogen™ Supplier No. MA523117
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MAASTASHRP IKGILKNKTS TTSSMVASAE QPRRSVDEEL SKKSQKWDEI NILATYHPAD KGYGLMKIDE PSPPYHSMMG DDEDACRDTE TTEAMAPGIL AKKLAAAEGL EPKYRIQEQE SSGEEDSDLS PEEREKKRQF EMRRKLHYNE GLNIKLARQL ISKDLHDDDE DEEMLETADG ESMNTEESNQ GSTPSDQQQN KLRSS.

Inhibitor of protein-phosphatase 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R2P3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1H2
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PPP1R2B
Gene Accession No. Q6NXS1
Gene Alias PPP1R2P3; protein phosphatase 1 regulatory inhibitor subunit 2 pseudogene 3
Gene Symbols PPP1R2B
Host Species Mouse
Immunogen PPP1R2P3 (AAH66922, 1 a.a. ~ 205 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153743
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.