Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R35 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564990
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564990 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564990 Supplier Invitrogen™ Supplier No. PA564990
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NAALREKLALL PPQARAPHPK EPPGPGPDMT ILCDPETLFY ESPHLTLDGL PPLRLQLRPR PSEDTFLMHR TLRRWEA Highest antigen sequence identity to the following orthologs - mouse 81%, rat 82%.

Inhibits PPP1CA phosphatase activity. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R35
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R35
Gene Accession No. Q8TAP8
Gene Alias 2010007H12Rik; 2010011D20Rik; AL024004; C7orf47; PPP1R35; Protein phosphatase 1 regulatory subunit 35; protein phosphatase 1, regulatory subunit 35; UPF0683 protein C7orf47; UPF0683 protein C7orf47 homolog
Gene Symbols PPP1R35
Host Species Rabbit
Immunogen Recombinant Human PPP1R35. Recombinant protein control fragment (Product #RP-92782).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221908
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.