Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139975
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139975 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139975 Supplier Invitrogen™ Supplier No. PA5139975
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: CNSTREIQGI EKHPYPESKP EEVSRSSGIV TSGSRKERCI GQIFQTEEYS VEKSLGPMIL INKPLENMEE ARHENEGLVS SGQSLYTSGE KESDSSASTS LPVEESQAQG NESLFS Highest antigen sequence indentity to the following orthologs: Mouse - 46%, Rat - 52%.

PPP1R3A, the glycogen-associated form of protein phosphatase-1 (PP1) derived from skeletal muscle is a heterodimer composed of a 37-kD catalytic subunit and a 124-kD targeting and regulatory subunit. PPP1R3A encodes the regulatory subunit which binds to muscle glycogen with high affinity, thereby enhancing dephosphorylation of glycogen-bound substrates for PP1 such as glycogen synthase and glycogen phosphorylase kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R3A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R3A
Gene Accession No. Q16821
Gene Alias glycogen and sarcoplasmic reticulum binding subunit, skeletal muscle; glycogen-associated regulatory subunit of protein phosphatase-1; GM; PP1G; PPP1R3; PPP1R3A; protein phosphatase 1 glycogen- associated regulatory subunit; protein phosphatase 1 glycogen-associated regulatory subunit; Protein phosphatase 1 regulatory subunit 3A; protein phosphatase 1 regulatory subunit GM; protein phosphatase 1, regulatory (inhibitor) subunit 3A; protein phosphatase 1, regulatory subunit 3A; protein phosphatase type-1 glycogen targeting subunit; RG1; serine /threonine specific protein phosphatase; type-1 protein phosphatase skeletal muscle glycogen targeting subunit
Gene Symbols PPP1R3A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP1R3A. Recombinant protein control fragment (Product #RP-109209).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5506
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.