Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP1R3B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP182243
Description
PPP1R3B Polyclonal specifically detects PPP1R3B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PPP1R3B | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| FLJ14005, FLJ34675, GL, Hepatic glycogen-targeting protein phosphatase 1 regulatory subunit GL, PPP1R4PP1 subunit R4, protein phosphatase 1 regulatory subunit 3B, Protein phosphatase 1 regulatory subunit 4, Protein phosphatase 1 subunit GL, protein phosphatase 1, regulatory (inhibitor) subunit 3B | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| PPP1R3B | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:DRDTFSFDISLPEKIQSYERMEFAVYYECNGQTYWDSNRGKNYRIIRAELKSTQGMTKPHSGPDLGISFDQFGSPRCSYGLF | |
| 0.1 mL | |
| metabolism | |
| 79660 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction