Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPP1R3B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107219
Description
Sequence homology: Cow: 93%; Dog: 93%; Goat: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%.
Acts as a glycogen-targeting subunit for phosphatase PP1. Facilitates interaction of the PP1 with enzymes of the glycogen metabolism and regulates its activity. Suppresses the rate at which PP1 dephosphorylates (inactivates) glycogen phosphorylase and enhances the rate at which it activates glycogen synthase and therefore limits glycogen breakdown. Its activity is inhibited by PYGL, resulting in inhibition of the glycogen synthase and glycogen phosphorylase phosphatase activities of PP1. Dramatically increases basal and insulin-stimulated glycogen synthesis upon overexpression in hepatocytes.
Specifications
PPP1R3B | |
Polyclonal | |
Unconjugated | |
PPP1R3B | |
33 kDa glycogen-binding protein; 6430576E21; AW821953; GL; Hepatic glycogen-targeting protein phosphatase 1 regulatory subunit GL; hepatic glycogen-targeting subunit, G(L); PP1 subunit R4; PPP1R3B; PPP1R4; protein phosphatase 1 (GL-subunit); protein phosphatase 1 regulatory subunit 3B; Protein phosphatase 1 regulatory subunit 4; protein phosphatase 1 subunit GL; protein phosphatase 1, regulatory (inhibitor) subunit 3B; protein phosphatase 1, regulatory subunit 3B; PTG; R4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79660 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
Q86XI6 | |
PPP1R3B | |
Synthetic peptide located within the following region: LSSKNEASGM VAPAVQEKKV KKRVSFADNQ GLALTMVKVF SEFDDPLDMP (aa 40-90). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction