Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555965
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555965 Supplier Invitrogen™ Supplier No. PA555965
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DRDTFSFDIS LPEKIQSYER MEFAVYYECN GQTYWDSNRG KNYRIIRAEL KSTQGMTKPH SGPDLGISFD QFGSPRCSYG LF Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 88%.

Acts as a glycogen-targeting subunit for phosphatase PP1. Facilitates interaction of the PP1 with enzymes of the glycogen metabolism and regulates its activity. Suppresses the rate at which PP1 dephosphorylates (inactivates) glycogen phosphorylase and enhances the rate at which it activates glycogen synthase and therefore limits glycogen breakdown. Its activity is inhibited by PYGL, resulting in inhibition of the glycogen synthase and glycogen phosphorylase phosphatase activities of PP1. Dramatically increases basal and insulin-stimulated glycogen synthesis upon overexpression in hepatocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R3B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R3B
Gene Accession No. Q86XI6
Gene Alias 33 kDa glycogen-binding protein; 6430576E21; AW821953; GL; Hepatic glycogen-targeting protein phosphatase 1 regulatory subunit GL; hepatic glycogen-targeting subunit, G(L); PP1 subunit R4; PPP1R3B; PPP1R4; protein phosphatase 1 (GL-subunit); protein phosphatase 1 regulatory subunit 3B; Protein phosphatase 1 regulatory subunit 4; protein phosphatase 1 subunit GL; protein phosphatase 1, regulatory (inhibitor) subunit 3B; protein phosphatase 1, regulatory subunit 3B; PTG; R4
Gene Symbols PPP1R3B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP1R3B. Recombinant protein control fragment (Product #RP-93907).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79660
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.