Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R3C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555493
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555493 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555493 Supplier Invitrogen™ Supplier No. PA555493
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PRPLTSSVMP VDVAMRLCLA HSPPVKSFLG PYDEFQRRHF VNKLKPLKSC LNIKHKAKSQ NDWKCSHNQA KKRVVFADSK GLSLTAIHVF SDLPEEPAWD LQFDLLDL Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 84%.

Protein phosphatase-1 (PP1; see MIM 176875) participates in the regulation of a wide variety of cellular functions by reversible protein phosphorylation. The ability of PP1 to regulate diverse functions resides in its capacity to interact with a variety of regulatory subunits that may target PP1 to specific subcellular locations, modulate its substrate specificity, and allow its activity to be responsive to extracellular signals. Several targeting subunits of PP1 have been identified, including PPP1R5, the glycogen-binding subunits PPP1R3 (MIM 600917) and PPP1R4, and the nuclear inhibitor of PP1 (PPP1R8; MIM 602636).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R3C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R3C
Gene Accession No. Q9UQK1
Gene Alias Phosphatase 1, regulatory inhibitor subunit 5; PP1 subunit R5; PPP1R3C; PPP1R5; protein phosphatase 1 binding protein PTG; protein phosphatase 1 regulatory subunit 3C; Protein phosphatase 1 regulatory subunit 5; protein phosphatase 1, regulatory (inhibitor) subunit 3C; protein phosphatase 1, regulatory (inhibitor) subunit 5; protein phosphatase 1, regulatory subunit 3C; protein targeting to glicogen; protein targeting to glycogen; PTG
Gene Symbols PPP1R3C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP1R3C. Recombinant protein control fragment (Product #RP-91067).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5507
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.