Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPP1R3D Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568285
Description
Predicted to react with Mouse and Rat samples.
Seems to act as a glycogen-targeting subunit for PP1. PP1 is essential for cell division, and participates in the regulation of glycogen metabolism, muscle contractility and protein synthesis.
Specifications
PPP1R3D | |
Polyclonal | |
Unconjugated | |
PPP1R3D | |
2610024M03Rik; Gm359; PP1 subunit R6; PPP1R3D; PPP1R6; protein phosphatase 1 regulatory subunit 3D; Protein phosphatase 1 regulatory subunit 6; protein phosphatase 1, regulatory (inhibitor) subunit 3D; protein phosphatase 1, regulatory subunit 3D; protein phosphatase 1, regulatory subunit 6, spinophilin; protein phosphatase 1-binding subunit R6 | |
Rabbit | |
affinity chromatography | |
RUO | |
228966, 5509, 689995 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O95685 | |
PPP1R3D | |
Synthesized peptide derived from human PPP1R3D. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction