Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R3D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559450
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559450 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559450 Supplier Invitrogen™ Supplier No. PA559450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELAQVKVFNA GDDPSVPLHV LSRLAINSDL CCSSQDLEFT LHCLVPDFPP PVEAADFGER LQRQLVCLER VTCSDL Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 89%.

Seems to act as a glycogen-targeting subunit for PP1. PP1 is essential for cell division, and participates in the regulation of glycogen metabolism, muscle contractility and protein synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R3D
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R3D
Gene Accession No. O95685
Gene Alias 2610024M03Rik; Gm359; PP1 subunit R6; PPP1R3D; PPP1R6; protein phosphatase 1 regulatory subunit 3D; Protein phosphatase 1 regulatory subunit 6; protein phosphatase 1, regulatory (inhibitor) subunit 3D; protein phosphatase 1, regulatory subunit 3D; protein phosphatase 1, regulatory subunit 6, spinophilin; protein phosphatase 1-binding subunit R6
Gene Symbols PPP1R3D
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP1R3D. Recombinant protein control fragment (Product #RP-99431).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5509
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.