Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R3E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563975
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563975 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563975 Supplier Invitrogen™ Supplier No. PA563975
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSRERPPGTD IPRNLSFIAA LTERAYYRSQ RPSLEEEPEE EP Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 90%.

Acts as a glycogen-targeting subunit for PP1. PP1 is involved in glycogen metabolism and contributes to the activation of glycogen synthase leading to an increase in glycogen synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R3E
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R3E
Gene Accession No. Q9H7J1
Gene Alias PPP1R3E; Protein phosphatase 1 regulatory subunit 3E; protein phosphatase 1, regulatory (inhibitor) subunit 3E; protein phosphatase 1, regulatory subunit 3E
Gene Symbols PPP1R3E
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP1R3E. Recombinant protein control fragment (Product #RP-102016).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90673
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.