Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R3F Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551485
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551485 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551485 Supplier Invitrogen™ Supplier No. PA551485
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TDGGMSPSHP LGILTDRDLI LKWPGPERAL NSALAEEITL HYARLGRGVE LIKDTEDPDD EGEGEEGLSV TPSSPEGDSP KESPPEILSG ARSVVATMGD VWLP Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 28%.

Glycogen-targeting subunit for protein phosphatase 1 (PP1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R3F
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R3F
Gene Accession No. Q6ZSY5
Gene Alias DXImx48e; HB2E; LL0XNC01-7P3.1; Ppp1r3f; Protein phosphatase 1 regulatory subunit 3F; protein phosphatase 1, regulatory (inhibitor) subunit 3F; protein phosphatase 1, regulatory subunit 3F; R3F; RF3; Sfc15
Gene Symbols PPP1R3F
Host Species Rabbit
Immunogen Recombinant protein fragment of Human PPP1R3F (Product #RP-91885)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89801
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.