Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPP1R3F Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118184
This item is not returnable.
View return policy
Description
PPP1R3F Polyclonal Antibody for Western Blot, IP
Glycogen-targeting subunit for protein phosphatase 1 (PP1).
Specifications
PPP1R3F | |
Polyclonal | |
PBS with 0.03% ProClin 300; pH 7 | |
Q6ZSY5 | |
PPP1R3F | |
E. coli-derived Human PPP1R3F fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
PPP1R3F | |
DXImx48e; HB2E; LL0XNC01-7P3.1; Ppp1r3f; Protein phosphatase 1 regulatory subunit 3F; protein phosphatase 1, regulatory (inhibitor) subunit 3F; protein phosphatase 1, regulatory subunit 3F; R3F; RF3; Sfc15 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
89801 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction