Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP1R3G Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571614
Description
Glycogen-targeting subunit for protein phosphatase 1 (PP1). Involved in the regulation of hepatic glycogenesis in a manner coupled to the fasting-feeding cycle and distinct from other glycogen-targeting subunits (By similarity).
Specifications
PPP1R3G | |
Polyclonal | |
Unconjugated | |
PPP1R3G | |
Protein phosphatase 1 regulatory subunit 3G; PPP1R3G | |
Rabbit | |
Protein A | |
RUO | |
648791 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.34 mg/mL | |
PBS with 0.09% sodium azide | |
B7ZBB8 | |
PPP1R3G | |
KLH conjugated synthetic peptide between 269-298 amino acids from the C-terminal region of human PPP1R3G | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction