Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R42 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555050
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555050 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555050 Supplier Invitrogen™ Supplier No. PA555050
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HSLAKSLCIL NISNNNIDDI TDLELLENLN QLIAVDNQLL HVKDLEFLLN KLMKLWKIDL NGNPVCLKPK YRDRLILVSK SLGT Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 67%.

Regulates phosphatase activity of protein phosphatase 1 (PP1) complexes in the testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R42
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R42
Gene Accession No. Q7Z4L9
Gene Alias 1700011J18Rik; 4930418G15Rik; dtr; leucine rich repeat containing 67; leucine-rich repeat-containing protein 67; Lrrc67; PPP1R42; Protein phosphatase 1 regulatory subunit 42; protein phosphatase 1, regulatory subunit 42; testis leucine-rich repeat; testis leucine-rich repeat protein; TLLR; TLRR
Gene Symbols PPP1R42
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP1R42. Recombinant protein control fragment (Product #RP-93967).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286187
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.