Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP1R7 Antibody (OTI4F9), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273586APC
Description
PPP1R7 Monoclonal antibody specifically detects PPP1R7 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
PPP1R7 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
OTI4F9 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
Protein phosphatase 1 regulatory subunit 22, protein phosphatase 1 regulatory subunit 7, protein phosphatase 1, regulatory (inhibitor) subunit 7, protein phosphatase-1 regulatory subunit 7 alpha2, protein phosphatase-1 regulatory subunit 7 beta1, protein phosphatase-1 regulatory subunit 7 beta2, regulatory subunit 7, sds22 | |
Full length human recombinant protein of human PPP1R7 (NP_002703) produced in HEK293T cell. | |
0.1 mL | |
Signal Transduction | |
5510 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction