Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556957
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556957 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556957 Supplier Invitrogen™ Supplier No. PA556957
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEHELPVDME TINLDRDAED VDLNHYRIGK IEGFEVLKKV KTLCLRQNLI KCIENLEELQ SLRELDLYDN QIKKIENL Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 36%.

In eukaryotes, the phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions, including division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the protein phosphatases. In general, the protein phosphatase 1 (PP1) holoenzyme is a trimeric complex composed of a regulatory subunit, a variable subunit, and a catalytic subunit. Sds22, also known as PPP1R7 (protein phosphatase 1, regulatory (inhibitor) subunit 7), is a 360 amino acid protein that localizes to the nucleus and contains ten LRR (leucine rich) repeats. Expressed in a variety of tissues, Sds22 functions as a regulatory subunit of the PP1 complex, suggesting a role in protein regulation throughout the cell. Multiple isoforms of Sds22 exist due to alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R7
Gene Accession No. Q15435
Gene Alias 2310014J01Rik; hypothetical protein LOC560323; im:7136431; Ppp1r7; Protein phosphatase 1 regulatory subunit 22; protein phosphatase 1 regulatory subunit 7; protein phosphatase 1, regulatory (inhibitor) subunit 7; protein phosphatase 1, regulatory subunit 7; protein phosphatase-1 regulatory subunit 7; Sds22; testis secretory sperm-binding protein Li 210a; zgc:123325
Gene Symbols PPP1R7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP1R7. Recombinant protein control fragment (Product #RP-96028).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5510
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.