Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPP1R8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586723
Description
Purity is > 95% by SDS-PAGE.
This gene, through alternative splicing, encodes three different isoforms. Two of the protein isoforms encoded by this gene are specific inhibitors of type 1 serine/threonine protein phosphatases and can bind but not cleave RNA. The third protein isoform lacks the phosphatase inhibitory function but is a single-strand endoribonuclease comparable to RNase E of E. coli. This isoform requires magnesium for its function and cleaves specific sites in A+U-rich regions of RNA.
Specifications
PPP1R8 | |
Polyclonal | |
Unconjugated | |
PPP1R8 | |
6330548N22Rik; activator of RNA decay; ARD1; ARD-1; AU044684; LOC100722083; NIPP1; NIPP-1; NIPP-1, nuclear inhibitor of protein phosphatase-1; Nuclear inhibitor of protein phosphatase 1; nuclear inhibitor of protein phosphatase-1 alpha; nuclear inhibitor of protein phosphatase-1 beta; nuclear subunit of PP-1; Ppp1r8; PRO2047; protein phosphatase 1 regulatory inhibitor subunit 8; protein phosphatase 1 regulatory subunit 8; protein phosphatase 1, regulatory (inhibitor) subunit 8; protein phosphatase 1, regulatory inhibitor subunit 8; protein phosphatase 1, regulatory subunit 8; RNase E | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100336, 313030, 5511 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q12972, Q8R3G1 | |
PPP1R8 | |
Synthetic peptide, corresponding to amino acids 237-282 of Human NIPP1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction