Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPP1R9A Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50
Specifications
Antigen | PPP1R9A |
---|---|
Concentration | 0.05 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5110944
|
Invitrogen™
PA5110944 |
100 μL |
Each for $589.50
|
|
|||||
PIPA5111734
|
Invitrogen™
PA5111734 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: KTKEGEGSQQ SRGRKYGSNV NRIKNLFMQM GMEPNENAAV IAKTRGKGGH SSPQRRMKPK EFLEKTDGSV VKLESSVSER IS.
Neurabin I is a brain-specific protein which contains an F-actin binding domain, a PDZ domain, a transmembrane-protein-interacting-domain, and a coiled-coil region. As its structure suggests, neurabin I is a promiscuous protein binding to F-actin, protein phosphatase 1, TGN38, and p70 S6 kinase. Found exclusively in brain, this protein is highly concentrated in the synapse and has also been shown to be enriched in the lamellipodia of the growth cone during neuronal development. Neurabin I appears to be a bridging protein by targeting other proteins to the synapse or by linking membrane proteins to the actin cytoskeleton.Specifications
PPP1R9A | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
PPP1R9A | |
2810430P21Rik; 4930518N04Rik; 5330407E15; A230094E16Rik; actin-binding protein neurabin; BB181831; KIAA1222; mKIAA1222; Neb1; neurabin 1; Neurabin I; neurabin-1; Neurabin-I; neural tissue-specific F-actin binding protein; neural tissue-specific F-actin-binding protein I; NRB; NRB1; NRBI; p180; PP1bp175; Ppp1r9a; Protein phosphatase 1 regulatory subunit 9A; protein phosphatase 1, regulatory (inhibitor) subunit 9A; protein phosphatase 1, regulatory subunit 9A | |
PPP1R9A | |
Primary | |
Antigen affinity chromatography |
0.05 mg/mL | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ULJ8 | |
55607 | |
Recombinant Protein corresonding to Human PPP1R9A. Recombinant protein control fragment (Product #RP-92313). | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title