Learn More
Invitrogen™ PPP1R9A Polyclonal Antibody

Description
Immunogen sequence: EEPCAESKAM PKSEIPSPQS QLLEDAEANL VGREAAKQQR KELAGGDFTS PDASASSCGK EVPEDSNNFD GSHVYMHSDY NVYRVRSRYN SDWGETGTEQ DEEEDSDENS YYQP Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 77%.
Specifications
Specifications
| Antigen | PPP1R9A |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | PPP1R9A |
| Gene Accession No. | Q9ULJ8 |
| Gene Alias | 2810430P21Rik; 4930518N04Rik; 5330407E15; A230094E16Rik; actin-binding protein neurabin; BB181831; KIAA1222; mKIAA1222; Neb1; neurabin 1; Neurabin I; neurabin-1; Neurabin-I; neural tissue-specific F-actin binding protein; neural tissue-specific F-actin-binding protein I; NRB; NRB1; NRBI; p180; PP1bp175; Ppp1r9a; Protein phosphatase 1 regulatory subunit 9A; protein phosphatase 1, regulatory (inhibitor) subunit 9A; protein phosphatase 1, regulatory subunit 9A |
| Gene Symbols | PPP1R9A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.