Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP1R9B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567692
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567692 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567692 Supplier Invitrogen™ Supplier No. PA567692
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

Protein phosphatase 1 regulatory subunit 9B (PPP1R9B), also known as Neurabin II or spinophilin, is a gene located on chromosome 17q21.32 that encodes a regulatory protein predominantly expressed in the brain. PPP1R9B is involved in modulating the activity of protein phosphatase 1 (PP1), a crucial enzyme in cellular signaling pathways responsible for dephosphorylation of target proteins. Spinophilin functions by anchoring PP1 to specific subcellular locations, such as synaptic sites, thereby influencing synaptic plasticity, neurotransmission, and signal transduction. This protein is integral to the maintenance and modulation of excitatory synapses, playing a role in cognitive processes including learning and memory. Alterations in PPP1R9B expression or function have been associated with neuropsychiatric disorders and cancer, reflecting its importance in neuronal integrity and its potential role in tumorigenesis. Research into PPP1R9B continues to explore its therapeutic potential in neurological disorders and its utility in understanding phosphatase-related cellular processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R9B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol, 0.87% NaCl and 0.01% sodium azide; pH 7.3
Gene Ppp1r9b
Gene Accession No. O35274, Q6R891, Q96SB3
Gene Alias Neb2; neurabin 2; neurabin II; neurabin-2; Neurabin-II; neural tissue-specific F-actin-binding protein II; nuerabin 2; p130; PP1bp134; PPP1R6; PPP1R9; PPP1R9B; Protein phosphatase 1 regulatory subunit 9B; protein phosphatase 1, regulatory (inhibitor) subunit 9B; protein phosphatase 1, regulatory subunit 9B; protein phosphatase 1, regulatory subunit 9B, spinophilin; SPINO; Spinophilin; SPL; Spn
Gene Symbols Ppp1r9b
Host Species Rabbit
Immunogen Synthesized peptide derived from human PPP1R9B.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217124, 84686, 84687
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.