Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP2CA Rabbit anti-Human, Rat, Unconjugated, Clone: AAA-16, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20557
Description
PPP2CA Monoclonal antibody specifically detects PPP2CA in Human, Rat samples. It is validated for IP, ICC, WB-Ce, IHC-P, IF.Specifications
PPP2CA | |
Monoclonal | |
Unconjugated | |
PPP2CA | |
Rabbit | |
100 μg | |
5515 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
AAA-16 | |
protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform | |
PP2Ac, PP2CA, RP-C | |
A synthetic phosphopeptide corresponding to residues surrounding Y307 of human PPP2CA | |
Primary | |
Human, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction