Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP2R1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674670
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-670 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-670 Supplier Invitrogen™ Supplier No. PA19086
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is expected to react with mouse, rat, canine, porcine, and bovine samples based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.

The PR65 subunit of protein phosphatase 2A serves as a scaffolding molecule to coordinate the assembly of the catalytic subunit and a variable regulatory B subunit. Upon interaction with GNA12 promotes dephosphorylation of microtubule associated protein TAU/MAPT. Required for proper chromosome segregation and for centromeric localization of SGO1 in mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R1A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene PPP2R1A
Gene Accession No. P30153
Gene Alias 29B; 6330556D22Rik; 65 kDa regulatory subunit of protein phosphatase; A/PR65; alpha isoform of regulatory subunit A, protein phosphatase 2; CG13383; CG17291; CG17291-PA; CG17291-PB; CG17291-PC; CG17291-PD; CG17291-PE; CG33297; Dmel\CG17291; Dmel_CG17291; dPP2A A; DPR65; Medium tumor antigen-associated 61 kDa protein; MGC786; MRD36; PP2A; PP2A 29B; PP2A 65kD subunit A; PP2A A; PP2A subunit A isoform PR65-alpha; PP2A subunit A isoform R1-alpha; PP2A[A]; Pp2A29B; PP2A-29B; Pp2A-29B-PA; Pp2A-29B-PB; Pp2A-29B-PC; Pp2A-29B-PD; Pp2A-29B-PE; PP2A-A; PP2AAALPHA; PP2A-Aalpha; PP2Ac; PP2CA; PP2Calpha; Ppp2r1a; PR65; PR65A; protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit A , alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit A, alpha isoform; protein phosphatase 2 regulatory subunit A, alpha; protein phosphatase 2 scaffold subunit Aalpha; protein phosphatase 2, regulatory subunit A, alpha; protein phosphatase 2A; protein phosphatase 2A at 29B; protein phosphatase PP2A; Protein phosphatase PP2A regulatory subunit A; protein phosphate 2A; regulatory subunit A of type 2A protein phosphatase; RP-C; serine/threonine protein phosphatase 2A, 65 kDa regulatory subunit A, alpha isoform; serine/threonine protein phosphatase A subunit type 2A; serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A alpha isoform; Serine/threonine-protein phosphatase PP2A 65 kDa regulatory subunit; testicular secretory protein Li 1; Unknown (protein for MGC:128399)
Gene Symbols PPP2R1A
Host Species Goat
Immunogen Synthetic peptide corresponding to the C terminus amino acids KYFAQEALTVLSLA (aa 576-589).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5518
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.