Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPP2R1A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103299
Description
Antibody detects endogenous levels of total PPP2R1A.
The PR65 subunit of protein phosphatase 2A serves as a scaffolding molecule to coordinate the assembly of the catalytic subunit and a variable regulatory B subunit. Upon interaction with GNA12 promotes dephosphorylation of microtubule associated protein TAU/MAPT. Required for proper chromosome segregation and for centromeric localization of SGO1 in mitosis.
Specifications
PPP2R1A | |
Polyclonal | |
Unconjugated | |
PPP2R1A | |
29B; 6330556D22Rik; 65 kDa regulatory subunit of protein phosphatase; A/PR65; alpha isoform of regulatory subunit A, protein phosphatase 2; CG13383; CG17291; CG17291-PA; CG17291-PB; CG17291-PC; CG17291-PD; CG17291-PE; CG33297; Dmel\CG17291; Dmel_CG17291; dPP2A A; DPR65; Medium tumor antigen-associated 61 kDa protein; MGC786; MRD36; PP2A; PP2A 29B; PP2A 65kD subunit A; PP2A A; PP2A subunit A isoform PR65-alpha; PP2A subunit A isoform R1-alpha; PP2A[A]; Pp2A29B; PP2A-29B; Pp2A-29B-PA; Pp2A-29B-PB; Pp2A-29B-PC; Pp2A-29B-PD; Pp2A-29B-PE; PP2A-A; PP2AAALPHA; PP2A-Aalpha; PP2Ac; PP2CA; PP2Calpha; Ppp2r1a; PR65; PR65A; protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit A , alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit A, alpha isoform; protein phosphatase 2 regulatory subunit A, alpha; protein phosphatase 2 scaffold subunit Aalpha; protein phosphatase 2, regulatory subunit A, alpha; protein phosphatase 2A; protein phosphatase 2A at 29B; protein phosphatase PP2A; Protein phosphatase PP2A regulatory subunit A; protein phosphate 2A; regulatory subunit A of type 2A protein phosphatase; RP-C; serine/threonine protein phosphatase 2A, 65 kDa regulatory subunit A, alpha isoform; serine/threonine protein phosphatase A subunit type 2A; serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A alpha isoform; Serine/threonine-protein phosphatase PP2A 65 kDa regulatory subunit; testicular secretory protein Li 1; Unknown (protein for MGC:128399) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
117281, 51792, 5518 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P30153, Q76MZ3 | |
PPP2R1A | |
A synthesized peptide derived from human PPP2R1A(Accession P30153), corresponding to amino acid residues L556-A589. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction