Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2R1A/PPP2R2A/PPP2R1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116861
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116861 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116861 Supplier Thermo Scientific Supplier No. PA5116861
Add to Cart
Add to Cart

PPP2R1A/PPP2R2A/PPP2R1B Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R1A/PPP2R2A/PPP2R1B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PPP2R1A
Gene Accession No. P30153, P30154, P36876, P63151, Q4QQT4, Q6P1F6, Q76MZ3, Q7TNP2
Gene Alias 2410004D02Rik; 2410091N08Rik; 6330556D22Rik; AI790395; alpha isoform of regulatory subunit A, protein phosphatase 2; B55A; B55ALPHA; Medium tumor antigen-associated 61 kDa protein; mKIAA1541; MRD36; PP2A; PP2A subunit A isoform PR65-alpha; PP2A subunit A isoform PR65-beta; PP2A subunit A isoform R1-alpha; PP2A subunit A isoform R1-beta; PP2A subunit B isoform alpha; PP2A subunit B isoform B55-alpha; PP2A subunit B isoform BRA; PP2A subunit B isoform PR55-alpha; PP2A subunit B isoform R2-alpha; PP2A, subunit A, PR65-beta isoform; PP2A, subunit A, R1-beta isoform; PP2A, subunit B, B55-alpha isoform; PP2A, subunit B, B-alpha isoform; PP2A, subunit B, BRA isoform; PP2A, subunit B, PR55-alpha isoform; PP2A, subunit B, R2-alpha isoform; PP2AAALPHA; PP2A-Aalpha; PP2A-Abeta; Ppp2r1a; Ppp2r1b; PPP2R2A; PR52A; PR55A; PR65; PR65A; PR65B; protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), beta isoform; protein phosphatase 2 (formerly 2A), regulatory subunit A, alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit A, beta isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR52), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B, alpha isoform; protein phosphatase 2 regulatory subunit A, alpha; protein phosphatase 2 regulatory subunit A, beta; protein phosphatase 2 regulatory subunit B, alpha; protein phosphatase 2 regulatory subunit Balpha; protein phosphatase 2 scaffold subunit Aalpha; protein phosphatase 2 scaffold subunit Abeta; protein phosphatase 2, regulatory subunit A, alpha; protein phosphatase 2, regulatory subunit A, beta; protein phosphatase 2, regulatory subunit B, alpha; protein phosphatase 2, structural/regulatory subunit A, beta; protein phosphatase PP2A; serine/threonine protein phosphatase 2A, 55 KDA regulatory subunit B, alpha isoform; serine/threonine protein phosphatase 2A, 65 kDa regulatory subunit A, alpha isoform; serine/threonine protein phosphatase A subunit type 2A; serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B alpha isoform; Serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A alpha isoform; serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A beta isoform; testicular secretory protein Li 1; testicular tissue protein Li 156
Gene Symbols PPP2R1A, PPP2R1B, PPP2R2A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-161 of human PPP2R1A/PPP2R2A/PPP2R1BPPP2R1A (NP_0550402).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117104, 117281, 315648, 51792, 5518, 5519, 5520, 71978, 73699
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.