Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2R1B Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP299604AF594

Catalog No. NB062013


Only null left
Add to Cart

Description

Description

PPP2R1B Polyclonal specifically detects PPP2R1B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

PPP2R1B
Polyclonal
Immunohistochemistry, Immunohistochemistry-Paraffin
MGC26454, PP2A subunit A isoform PR65-beta, PP2A subunit A isoform R1-beta, PP2A, subunit A, PR65-beta isoform, PP2A, subunit A, R1-beta isoform, PP2A-Abeta, PR65B, protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), beta isoform, protein phosphatase 2 (formerly 2A), regulatory subunit A, beta isoform, protein phosphatase 2, regulatory subunit A, beta, protein phosphatase 2, structural/regulatory subunit A, beta, serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A betaisoform
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human PPP2R1B.
0.1 mL
Breast Cancer, Protein Phosphatase, Wnt Signaling Pathway
5519
Store at 4°C in the dark.
IgG
Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 594
50mM Sodium Borate
Rabbit
Antigen Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.