Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP2R1B Monoclonal Antibody (OTI9F3)
GREENER_CHOICE

Catalog No. PIMA527455
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527455 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527455 Supplier Invitrogen™ Supplier No. MA527455
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PPP2R1B Monoclonal Antibody for Western Blot

In eukaryotes, the phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions, including division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the protein phosphatases. In general, the protein phosphatase (PP) holoenzyme is a trimeric complex composed of a regulatory subunit, a variable subunit and a catalytic subunit. Four major families of protein phosphatase catalytic subunits have been identified, designated PP1, PP2A, PP2B (calcineurin) and PP2C. An additional protein phosphatase catalytic subunit, PPX (also known as PP4), is a putative member of a novel PP family. The PP2A family comprises subfamily members PP2A alpha and PP2A beta. The PP2A catalytic subunit associates with a variety of regulatory subunits. Regulatory subunits include PP2A-A alpha and -A beta, PP2A-B alpha and -B beta, PP2A-C alpha and -C beta, and PP2A-B56 alpha and -B56 beta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R1B
Applications Western Blot
Classification Monoclonal
Clone OTI9F3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 1% BSA and 0.02% sodium azide; pH 7.3
Gene Ppp2r1b
Gene Accession No. P30154
Gene Alias 2410091N08Rik; AI790395; PP2A A beta; PP2A -AR1 beta; PP2A subunit A isoform PR65-beta; PP2A subunit A isoform R1-beta; PP2A, subunit A, PR65-beta isoform; PP2A, subunit A, R1-beta isoform; PP2AA beta; PP2A-A beta; PP2A-Abeta; PP2A-APR65 beta; PPP2R1B; PR65B; protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), beta isoform; protein phosphatase 2 (formerly 2A), regulatory subunit A, beta isoform; protein phosphatase 2 regulatory subunit A, beta; protein phosphatase 2 scaffold subunit Abeta; protein phosphatase 2, regulatory subunit A, beta; protein phosphatase 2, structural/regulatory subunit A, beta; serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A beta isoform
Gene Symbols Ppp2r1b
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 19-601 of PPP2R1B produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5519
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.