Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP2R2A Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306295AF350
Description
PPP2R2A Polyclonal specifically detects PPP2R2A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PPP2R2A | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| B55A, B55ALPHA, DKFZp686N05117, FLJ26613, FLJ41727, MGC52248, PP2A subunit B isoform alpha, PP2A subunit B isoform B55-alpha, PP2A subunit B isoform PR55-alpha, PP2A subunit B isoform R2-alpha, PR52A, PR55A, protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), alphaisoform, protein phosphatase 2 (formerly 2A), regulatory subunit B (PR52), alpha isoform, protein phosphatase 2 (formerly 2A), regulatory subunit B, alpha isoform, protein phosphatase 2, regulatory subunit B, alpha, serine/threonine protein phosphatase 2A, 55 KDA regulatory subunit B, alphaisoform, serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B alphaisoform | |
| Produced in rabbits immunized with E. coli-derived Human PPP2R2A fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 5520 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction