Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPP2R2A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116564
Description
Positive Samples: HeLa, Jurkat, Mouse brain Immunogen sequence: RWLPQKNAAQ FLLSTNDKTI KLWKISERDK RPEGYNLKEE DGRYRDPTTV TTLRVPVFRP MDLMVEASPR RIFANAHTYH INSISINSDY ETYLSADDLR I.
The product of this gene belongs to the phosphatase 2 regulatory subunit B family. Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes an alpha isoform of the regulatory subunit B55 subfamily. Alternatively spliced transcript variants have been described.
Specifications
PPP2R2A | |
Polyclonal | |
Unconjugated | |
PPP2R2A | |
2410004D02Rik; B55A; B55ALPHA; hypothetical protein; mKIAA1541; PP2A subunit B isoform alpha; PP2A subunit B isoform B55-alpha; PP2A subunit B isoform BRA; PP2A subunit B isoform PR55-alpha; PP2A subunit B isoform R2-alpha; PP2A, subunit B, B55-alpha isoform; PP2A, subunit B, B-alpha isoform; PP2A, subunit B, BRA isoform; PP2A, subunit B, PR55-alpha isoform; PP2A, subunit B, R2-alpha isoform; PPP2R2A; PR52A; PR55A; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR52), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B, alpha isoform; protein phosphatase 2 regulatory subunit B, alpha; protein phosphatase 2 regulatory subunit Balpha; protein phosphatase 2, regulatory subunit B, alpha; protein phosphatase 2A 55 kDa regulatory subunit, alpha isoform; serine/threonine protein phosphatase 2A, 55 KDA regulatory subunit B, alpha isoform; serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B alpha isoform; serine-threonine protein phosphatase 2A regulatory subunit; testicular tissue protein Li 156 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
5520, 71978 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.71 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P63151, Q6P1F6 | |
PPP2R2A | |
Recombinant fusion protein containing a sequence corresponding to amino acids 100-200 of human PP2A-B55a/PR55a/PPP2R2A (NP_0027081). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction