Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP2R2A/PPP2R2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674124
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-124 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-124 Supplier Invitrogen™ Supplier No. PA112391
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

PPP2R2A/PPP2R2B Polyclonal Antibody for Western Blot

The product of this gene belongs to the phosphatase 2 regulatory subunit B family. Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes a beta isoform of the regulatory subunit B55 subfamily. Defects in this gene cause autosomal dominant spinocerebellar ataxia 12 (SCA12), a disease caused by degeneration of the cerebellum, sometimes involving the brainstem and spinal cord, and in resulting in poor coordination of speech and body movements. Multiple alternatively spliced variants, which encode different isoforms, have been identified for this gene. The 5' UTR of some of these variants includes a CAG trinucleotide repeat sequence (7-28 copies) that can be expanded to 66-78 copies in cases of SCA12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R2A/PPP2R2B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene PPP2R2A
Gene Accession No. P36876, P36877, P63151, Q00005, Q5E9Q7, Q6P1F6, Q6ZWR4
Gene Alias 2410004D02Rik; 2900026H06Rik; 6330404L05Rik; B55A; B55ALPHA; B55BETA; beta isoform of regulatory subunit B55, protein phosphatase 2 isoform a; BRbeta B-regulatory subunit of protein phosphatase 2A; E130009M08Rik; mKIAA1541; PP2A subunit B isoform alpha; PP2A subunit B isoform B55-alpha; PP2A subunit B isoform B55-beta; PP2A subunit B isoform beta; PP2A subunit B isoform BRA; PP2A subunit B isoform BRB; PP2A subunit B isoform PR55-alpha; PP2A subunit B isoform PR55-beta; PP2A subunit B isoform R2-alpha; PP2A subunit B isoform R2-beta; PP2A, subunit B, B55-alpha isoform; PP2A, subunit B, B55-beta isoform; PP2A, subunit B, B-alpha isoform; PP2A, subunit B, B-beta isoform; PP2A, subunit B, BRA isoform; PP2A, subunit B, BRB isoform; PP2A, subunit B, PR55-alpha isoform; PP2A, subunit B, PR55-beta isoform; PP2A, subunit B, R2-alpha isoform; PP2A, subunit B, R2-beta isoform; PP2AB55BETA; PP2ABBETA; PP2APR55B; PP2A-PR55B; PP2APR55BETA; PPP2R2A; Ppp2r2b; Pppr2b2; PR2AB55BETA; PR2ABBETA; PR2APR55BETA; PR52A; PR52B; PR55A; PR55BETA; PR55-BETA; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), beta isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR52), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B, alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B, beta isoform; protein phosphatase 2 regulatory subunit B, alpha; protein phosphatase 2 regulatory subunit B, beta; protein phosphatase 2 regulatory subunit Balpha; protein phosphatase 2 regulatory subunit Bbeta; protein phosphatase 2, regulatory subunit B, alpha; protein phosphatase 2, regulatory subunit B, beta; SCA12; serine/threonine protein phosphatase 2A, 55 KDA regulatory subunit B, alpha isoform; serine/threonine protein phosphatase 2A, 55 kDa regulatory subunit B, beta isoform; serine/threonine protein phosphatase 2A, neuronal isoform; serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B alpha isoform; Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B beta isoform; testicular tissue protein Li 156
Gene Symbols PPP2R2A, PPP2R2B
Host Species Sheep
Immunogen Purified peptide conjugated to KLH.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117104, 508045, 509290, 5520, 5521, 60660, 71978, 72930
Target Species Human, Mouse, Rat, Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.