Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP2R2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587943
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587943 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587943 Supplier Invitrogen™ Supplier No. PA587943
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KIRWLPQQNA AYFLLSTNDK TVKLWKVSER DKRPEGYNLK DEEGRLRDPA TITTLRVPVL RPMDLMVEAT PRRVFANAHT YHINSISVNS DYETYMSADD L; Positive Samples: U-87MG, SH-SY5Y, HepG2, Mouse brain, Mouse lung, Rat brain; Cellular Location: Cytoplasm, Membrane, Mitochondrion, Mitochondrion outer membrane, cytoskeleton.

The product of this gene belongs to the phosphatase 2 regulatory subunit B family. Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes a beta isoform of the regulatory subunit B55 subfamily. Defects in this gene cause autosomal dominant spinocerebellar ataxia 12 (SCA12), a disease caused by degeneration of the cerebellum, sometimes involving the brainstem and spinal cord, and in resulting in poor coordination of speech and body movements. Multiple alternatively spliced variants, which encode different isoforms, have been identified for this gene. The 5' UTR of some of these variants includes a CAG trinucleotide repeat sequence (7-28 copies) that can be expanded to 66-78 copies in cases of SCA12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R2B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PPP2R2B
Gene Accession No. P36877, Q00005, Q6ZWR4
Gene Alias 2900026H06Rik; 6330404L05Rik; B55BETA; beta isoform of regulatory subunit B55, protein phosphatase 2 isoform a; BRbeta B-regulatory subunit of protein phosphatase 2A; E130009M08Rik; PP2A PP2ABBETA; PP2A subunit B isoform B55-beta; PP2A subunit B isoform beta; PP2A subunit B isoform BRB; PP2A subunit B isoform PR55-beta; PP2A subunit B isoform R2-beta; PP2A, subunit B, B55-beta isoform; PP2A, subunit B, B-beta isoform; PP2A, subunit B, BRB isoform; PP2A, subunit B, PR55-beta isoform; PP2A, subunit B, R2-beta isoform; PP2AB55BETA; PP2ABBETA; PP2APR55B; PP2A-PR55B; PP2APR55BETA; PPP2R2B; Pppr2b2; PR2A PR2ABBETA; PR2AB55BETA; PR2ABBETA; PR2APR55BETA; PR52B; PR55BETA; PR55-BETA; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), beta isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B, beta isoform; protein phosphatase 2 regulatory subunit B, beta; protein phosphatase 2 regulatory subunit Bbeta; protein phosphatase 2, regulatory subunit B, beta; SCA12; serine/threonine protein phosphatase 2A, 55 kDa regulatory subunit B, beta isoform; serine/threonine protein phosphatase 2A, neuronal isoform; serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B beta isoform
Gene Symbols PPP2R2B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 160-260 of human PP2A-B55β/PR55β/PP2A-B55β/PR55β/PPP2R2B (NP_8580602).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5521, 60660, 72930
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.