Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP2R2B/PPP2R2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116862
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116862 Supplier Invitrogen™ Supplier No. PA5116862
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: MCF7, Mouse brain, Mouse testis, Rat brain, Rat testis Immunogen sequence: GDKGGRVVIF QREQESKNQV HRRGEYNVYS TFQSHEPEFD YLKSLEIEEK INKIRWLPQQ NAAYFLLSTN DKTVKLWKVS ERDKRPEGYN LKDEEGRLRD PATITTLRVP VLR.

Protein phosphatase 3 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The constant regulatory subunit A serves as a scaffolding molecule to coordinate the assembly of the catalytic subunit and a variable regulatory B subunit.The B regulatory subunit might modulate substrate selectivity and catalytic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R2B/PPP2R2C
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PPP2R2C
Gene Accession No. P36877, P97888, Q00005, Q6ZWR4, Q8BG02, Q9Y2T4
Gene Alias 2900026H06Rik; 6330404L05Rik; 6330548O06Rik; B55BETA; B55-GAMMA; BRbeta B-regulatory subunit of protein phosphatase 2A; E130009M08Rik; gamma isoform of regulatory subunit B55, protein phosphatase 2; IMYPNO; IMYPNO1; phosphoprotein phosphatase 2A BR gamma regulatory chain; PP2A subunit B isoform B55-beta; PP2A subunit B isoform B55-gamma; PP2A subunit B isoform beta; PP2A subunit B isoform BRB; PP2A subunit B isoform BRgamma; PP2A subunit B isoform gamma; PP2A subunit B isoform PR55-beta; PP2A subunit B isoform PR55-gamma; PP2A subunit B isoform R2-beta; PP2A subunit B isoform R2-gamma; PP2A, subunit B, B55-beta isoform; PP2A, subunit B, B55-gamma isoform; PP2A, subunit B, B-beta isoform; PP2A, subunit B, B-gamma isoform; PP2A, subunit B, BRB isoform; PP2A, subunit B, BRgamma isoform; PP2A, subunit B, PR55-beta isoform; PP2A, subunit B, PR55-gamma isoform; PP2A, subunit B, R2-beta isoform; PP2A, subunit B, R2-gamma isoform; PP2AB55BETA; PP2ABBETA; PP2APR55B; PP2A-PR55B; PP2APR55BETA; PPP2R2B; Ppp2r2c; Pppr2b2; PR2AB55BETA; PR2ABBETA; PR2APR55BETA; PR52; PR52B; PR55BETA; PR55-BETA; PR55G; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), beta isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), gamma isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B, beta isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B, gamma isoform; protein phosphatase 2 regulatory subunit B, beta; protein phosphatase 2 regulatory subunit B, gamma; protein phosphatase 2 regulatory subunit Bbeta; protein phosphatase 2 regulatory subunit Bgamma; protein phosphatase 2, regulatory subunit B, beta; protein phosphatase 2, regulatory subunit B, gamma; protein phosphatase 2A1 B gamma subunit; SCA12; serine/threonine protein phosphatase 2A, 55 kDa regulatory subunit B, beta isoform; serine/threonine protein phosphatase 2A, neuronal isoform; serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B beta isoform; Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B gamma isoform
Gene Symbols PPP2R2B, PPP2R2C
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 108-220 of human PPP2R2B/PPP2R2CPPP2R2B (NP_8580602).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117256, 269643, 5521, 5522, 60660, 72930
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.