Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPP2R2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549389
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549389 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549389 Supplier Invitrogen™ Supplier No. PA549389
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with monkey, mouse, rabbit and rat based on sequence homology.

The B regulatory subunit might modulate substrate selectivity and catalytic activity, and also might direct the localization of the catalytic enzyme to a particular subcellular compartment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP2R2C
Applications Western Blot
Classification Polyclonal
Concentration mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PPP2R2C
Gene Accession No. Q9Y2T4
Gene Alias 6330548O06Rik; B55-GAMMA; gamma isoform of regulatory subunit B55, protein phosphatase 2; IMYPNO; IMYPNO1; phosphoprotein phosphatase 2A BR gamma regulatory chain; PP2A subunit B isoform B55-gamma; PP2A subunit B isoform BRgamma; PP2A subunit B isoform gamma; PP2A subunit B isoform PR55-gamma; PP2A subunit B isoform R2-gamma; PP2A, subunit B, B55-gamma isoform; PP2A, subunit B, B-gamma isoform; PP2A, subunit B, BRgamma isoform; PP2A, subunit B, PR55-gamma isoform; PP2A, subunit B, R2-gamma isoform; Ppp2r2c; Pr52; PR55G; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), gamma isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B, gamma isoform; protein phosphatase 2 regulatory subunit B, gamma; protein phosphatase 2 regulatory subunit Bgamma; protein phosphatase 2, regulatory subunit B, gamma; protein phosphatase 2A1 B gamma subunit; serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B gamma isoform
Gene Symbols PPP2R2C
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 53-86 amino acids from the N-terminal region of human PPP2R2C.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5522
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.